Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015982.3 | NP_057066.2 | 9 | 1291 | ||
XM_017024713.1 | XP_016880202.1 | 10 | 1381 | |||
GenBank mRNA | AB208818.1 | - | 6 | 4272 | ||
AF096834.1 | - | 9 | 1289 | |||
AK092747.1 | - | 8 | 1908 | |||
BC033800.1 | - | 9 | 1254 | 5171696 | ||
BC047760.1 | - | 9 | 1255 | 5742121 |