Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLLP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015993.2 | NP_057077.1 | 2 | 343 | ||
GenBank mRNA | AF137386.1 | - | 2 | 339 | ||
AK130584.1 | - | 2 | 335 | |||
AK289599.1 | - | 2 | 320 | |||
AK313910.1 | - | 2 | 343 | |||
BC002760.2 | - | 2 | 306 | 3633345 | ||
DA366260.1 | - | 2 | 344 |