Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321102.1 | NP_001308031.1 | 3 | 1277 | ||
NM_016030.5 | NP_057114.5 | 3 | 1314 | |||
XM_011510350.2 | XP_011508652.1 | 3 | 1744 | |||
XM_011510352.2 | XP_011508654.1 | 3 | 1744 | |||
XM_011510353.2 | XP_011508655.1 | 3 | 1744 | |||
XM_011510354.2 | XP_011508656.1 | 3 | 1744 | |||
XM_011510355.2 | XP_011508657.1 | 2 | 255 | |||
XM_011510356.1 | XP_011508658.1 | 2 | 147 | |||
XM_017004257.1 | XP_016859746.1 | 3 | 1744 | |||
GenBank mRNA | AF151845.1 | - | 1 | 43 | ||
AK094242.1 | - | 2 | 688 | |||
AK098327.1 | - | 3 | 1279 | |||
BC014164.2 | - | 3 | 1191 | 4548681 | ||
BC017475.1 | - | 2 | 312 |