Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 5 | 1054 | ||
NM_016032.3 | NP_057116.2 | 6 | 1034 | |||
GenBank mRNA | AB209807.1 | - | 5 | 973 | ||
AF151847.1 | - | 6 | 922 | |||
AK001112.1 | - | 6 | 984 | |||
AK001424.1 | - | 3 | 368 | |||
AK001524.1 | - | 5 | 785 | |||
AK096967.1 | - | 6 | 685 | |||
AL161962.1 | - | 6 | 911 | |||
AY358558.1 | - | 6 | 966 | |||
AY952881.8 | - | 6 | 1006 | |||
BC000035.2 | - | 5 | 928 | 3506601 | ||
BC003128.1 | - | 6 | 972 | 2957911 | ||
BC006200.2 | - | 6 | 968 | 2964425 | ||
BC012826.2 | - | 6 | 968 | 2964425 |