Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008222.2 | NP_001008223.1 | 10 | 1456 | ||
NM_016032.3 | NP_057116.2 | 11 | 1436 | |||
GenBank mRNA | AB209807.1 | - | 10 | 1375 | ||
AF151847.1 | - | 11 | 1324 | |||
AK001112.1 | - | 11 | 1386 | |||
AK001424.1 | - | 8 | 770 | |||
AK001524.1 | - | 10 | 1187 | |||
AK096967.1 | - | 11 | 1087 | |||
AL161962.1 | - | 11 | 1313 | |||
AY358558.1 | - | 11 | 1368 | |||
AY952881.8 | - | 11 | 1408 | |||
BC000035.2 | - | 10 | 1330 | 3506601 | ||
BC003128.1 | - | 11 | 1374 | 2957911 | ||
BC006200.2 | - | 11 | 1370 | 2964425 | ||
BC012826.2 | - | 11 | 1370 | 2964425 |