Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC25B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287516.1 | NP_001274445.1 | 11 | 1345 | ||
NM_001287517.1 | NP_001274446.1 | 11 | 1303 | |||
NM_001287518.1 | NP_001274447.1 | 10 | 1264 | |||
NM_001287519.1 | NP_001274448.1 | 11 | 1076 | |||
NM_001287520.1 | NP_001274449.1 | 11 | 1118 | |||
NM_001287522.1 | NP_001274451.1 | 10 | 995 | |||
NM_001287524.1 | NP_001274453.1 | 7 | 900 | |||
NM_004358.4 | NP_004349.1 | 11 | 1928 | |||
NM_021872.3 | NP_068658.1 | 10 | 1847 | |||
NM_021873.3 | NP_068659.1 | 11 | 1970 | |||
NR_136335.1 | 11 | 1118 | ||||
NR_136336.1 | 11 | 1345 | ||||
GenBank mRNA | AK092713.1 | - | 11 | 1118 | ||
AK295573.1 | - | 11 | 1187 | |||
AK299028.1 | - | 10 | 1071 | |||
AK299192.1 | - | 11 | 1121 | |||
BC006395.2 | - | 11 | 1251 | 4135465 | ||
BC009953.2 | - | 11 | 1331 | 4130804 | ||
BC051711.1 | - | 11 | 1875 | 5301205 | ||
BX640836.1 | - | 7 | 892 | |||
BX647988.1 | - | 11 | 1033 | |||
FR695900.1 | - | 11 | 1118 | |||
FR695901.1 | - | 10 | 995 | |||
M81934.1 | - | 11 | 1207 | |||
S78187.1 | - | 11 | 1375 | |||
Z68092.1 | - | 10 | 1155 |