Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016216.3 | NP_057300.2 | 5 | 723 | ||
GenBank mRNA | AF180919.2 | - | 5 | 677 | ||
AK094147.1 | - | 6 | 608 | |||
AL702928.1 | - | 5 | 725 | |||
BC009472.1 | - | 5 | 651 | 3503627 |