Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCTN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135643.1 | NP_001129115.1 | 13, 14 | 1289 | ||
NM_001135644.1 | NP_001129116.1 | 12, 13 | 1436 | |||
NM_016221.3 | NP_057305.1 | 12, 13 | 1268 | |||
XM_005268457.1 | XP_005268514.1 | 11, 12 | 1162 | |||
XM_011537644.1 | XP_011535946.1 | 13, 14 | 1172 | |||
XM_011537645.1 | XP_011535947.1 | 13, 14 | 1084 | |||
XM_011537646.1 | XP_011535948.1 | 13, 14 | 1305 | |||
XM_011537647.1 | XP_011535949.1 | 13, 14 | 1128 | |||
GenBank mRNA | AF195120.1 | - | 12, 13 | 1183 | ||
AK000299.1 | - | 12, 13 | 1187 | |||
AK125973.1 | - | 12, 13 | 1436 | |||
AK296787.1 | - | 4, 5 | 448 | |||
AK299517.1 | - | 12, 13 | 1189 | |||
AK307757.1 | - | 13, 14 | 1228 | |||
AK311693.1 | - | 9, 10 | 984 | |||
AL162085.1 | - | 2, 3 | 218 | |||
BC026323.1 | - | 12, 13 | 1193 | 4815273 | ||
KU178637.1 | - | 12, 13 | 1166 | |||
KU178638.1 | - | 12, 13 | 1109 | |||
KU178639.1 | - | 8, 9 | 922 |