Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164372.1 | NP_001157844.1 | 7 | 896 | ||
NM_001164373.1 | NP_001157845.1 | 7 | 805 | |||
NM_016301.3 | NP_057385.3 | 7 | 775 | |||
XM_005253896.4 | XP_005253953.3 | 8 | 1230 | |||
XM_017019394.1 | XP_016874883.1 | 7 | 710 | |||
GenBank mRNA | AF117229.1 | - | 7 | 725 | ||
AK223209.1 | - | 7 | 747 | |||
AK314946.1 | - | 7 | 748 | |||
AM992619.1 | - | 7 | 694 | |||
AM992620.1 | - | 7 | 694 | |||
AY359078.1 | - | 7 | 776 | |||
AY364262.1 | - | 7 | 694 | |||
BC008416.1 | - | 7 | 750 | 4048678 | ||
BC031024.1 | - | 7 | 751 | 4695834 | ||
BX537973.1 | - | 7 | 786 |