Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323625.1 | NP_001310554.1 | 4 | 992 | ||
NM_016361.4 | NP_057445.4 | 4 | 961 | |||
NR_136633.1 | 4 | 992 | ||||
NR_136634.1 | 4 | 1312 | ||||
NR_136635.1 | 4 | 992 | ||||
NR_136636.1 | 4 | 1312 | ||||
XM_011509601.2 | XP_011507903.1 | 4 | 917 | |||
XM_017001422.1 | XP_016856911.1 | 4 | 917 | |||
GenBank mRNA | AB030039.2 | - | 4 | 765 | ||
AB031478.1 | - | 4 | 872 | |||
AB209248.1 | - | 4 | 734 | |||
AK000657.1 | - | 4 | 698 | |||
AK308593.1 | - | 3 | 599 | |||
AK310693.1 | - | 2 | 468 | |||
AY358552.1 | - | 4 | 669 | |||
BC009965.1 | - | 4 | 941 | 4299943 | ||
BC034686.1 | - | 4 | 945 | 3463770 | ||
CR457101.1 | - | 4 | 476 | |||
KJ534759.1 | - | 4 | 497 | |||
KJ535036.1 | - | 3 | 369 | |||
KJ535044.1 | - | 4 | 497 | |||
KJ535055.1 | - | 4 | 497 |