Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016436.4 | NP_057520.2 | 10 | 1453 | ||
XM_005260421.2 | XP_005260478.1 | 7 | 1063 | |||
XM_011528843.1 | XP_011527145.1 | 9 | 1054 | |||
XM_011528844.1 | XP_011527146.1 | 7 | 1125 | |||
XM_017027864.1 | XP_016883353.1 | 11 | 1535 | |||
XM_017027865.1 | XP_016883354.1 | 11 | 1736 | |||
XM_017027866.1 | XP_016883355.1 | 10 | 1656 | |||
XM_017027867.1 | XP_016883356.1 | 10 | 1491 | |||
XM_017027868.1 | XP_016883357.1 | 8 | 1277 | |||
XM_017027869.1 | XP_016883358.1 | 10 | 1450 | |||
XM_017027870.1 | XP_016883359.1 | 10 | 1456 | |||
XM_017027871.1 | XP_016883360.1 | 11 | 1536 | |||
GenBank mRNA | AF220416.1 | - | 11 | 1356 | ||
AF258787.1 | - | 10 | 1439 | |||
AF348207.1 | - | 9 | 1378 | |||
AK054597.1 | - | 11 | 1710 | |||
AK314503.1 | - | 10 | 1417 | |||
AY027523.1 | - | 9 | 1378 | |||
AY134747.1 | - | 10 | 1430 | |||
BC006415.1 | - | 9 | 1313 | |||
BC016948.1 | - | 10 | 1406 | |||
BC048210.1 | - | 11 | 1486 | |||
BC080598.1 | - | 10 | 1417 | |||
BC093405.1 | - | 10 | 1453 | |||
BC150178.1 | - | 10 | 1419 | 40148074 | ||
L09749.1 | - | 9 | 1333 |