Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272065.1 | NP_001258994.1 | 2 | 310 | ||
NM_001272066.1 | NP_001258995.1 | 2 | 276 | |||
NM_001272067.1 | NP_001258996.1 | 2 | 471 | |||
NM_001272068.1 | NP_001258997.1 | 3 | 508 | |||
NM_016479.4 | NP_057563.3 | 2 | 310 | |||
XM_005265205.1 | XP_005265262.1 | 3 | 558 | |||
XM_006713188.2 | XP_006713251.1 | 2 | 211 | |||
XM_011533796.1 | XP_011532098.1 | 3 | 300 | |||
GenBank mRNA | AF520698.1 | - | 2 | 289 | ||
AK056328.1 | - | 2 | 452 | |||
AK075441.1 | - | 2 | 225 | |||
AK124652.1 | - | 3 | 508 | |||
BI552637.1 | - | 2 | 315 | |||
BX336842.2 | - | 2 | 193 | |||
BX344125.2 | - | 2 | 237 | |||
BX379507.2 | - | 2 | 202 | |||
DA009534.1 | - | 2 | 218 | |||
DA020280.1 | - | 2 | 471 | |||
DA025113.1 | - | 2 | 257 | |||
DA045437.1 | - | 2 | 276 | |||
DA657994.1 | - | 2 | 263 |