Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDKL3 SIRNA Assays ›
Gene Symbol:
CDKL3
Gene Name:
cyclin dependent kinase like 3
Gene Aliases:
NKIAMRE
Chromosome Location:
Chr.5: 134298424 - 134371159 on Build GRCh38
UniGene ID:
Hs.719926
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27797
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDKL3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113575.1
NP_001107047.1
9
1350
NM_001300853.1
NP_001287782.1
8
1116
NM_016508.3
NP_057592.2
9
1350
XM_011543429.2
XP_011541731.1
9
1701
XM_011543439.2
XP_011541741.1
6
854
XM_011543444.2
XP_011541746.1
5
683
XM_011543445.2
XP_011541747.1
5
632
XM_011543447.2
XP_011541749.1
4
543
XM_017009524.1
XP_016865013.1
9
2178
XM_017009525.1
XP_016865014.1
10
1746
XM_017009526.1
XP_016865015.1
10
1744
XM_017009527.1
XP_016865016.1
9
2142
XM_017009528.1
XP_016865017.1
8
2133
XM_017009529.1
XP_016865018.1
9
2178
XM_017009530.1
XP_016865019.1
9
2178
XM_017009531.1
XP_016865020.1
9
2178
XM_017009532.1
XP_016865021.1
8
2096
XM_017009533.1
XP_016865022.1
8
2132
XM_017009534.1
XP_016865023.1
8
2132
XM_017009535.1
XP_016865024.1
8
2096
XM_017009536.1
XP_016865025.1
6
1891
XM_017009537.1
XP_016865026.1
7
1296
XM_017009538.1
XP_016865027.1
9
1186
XM_017009539.1
XP_016865028.1
8
1007
XM_017009540.1
XP_016865029.1
7
817
XM_017009541.1
XP_016865030.1
8
1343
XM_017009542.1
XP_016865031.1
7
774
XM_017009543.1
XP_016865032.1
7
801
XM_017009544.1
XP_016865033.1
7
1052
XM_017009545.1
XP_016865034.1
6
859
XM_017009546.1
XP_016865035.1
6
888
XM_017009547.1
XP_016865036.1
4
580
XM_017009548.1
XP_016865037.1
7
1983
GenBank mRNA
AF130372.1
-
8
1231
AK294559.1
-
6
738
AK294740.1
-
5
648
AK297586.1
-
4
510
AK299305.1
-
5
598
AK301802.1
-
8
1110
AK310955.1
-
5
588
BC041799.1
-
9
1352
5298279
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein phosphorylation
regulation of cell cycle
protein kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-jmz56:80/10.239.163.191:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0