Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GMPR2 SIRNA Assays ›
Gene Symbol:
GMPR2
Gene Name:
guanosine monophosphate reductase 2
Gene Aliases:
GMPR 2
Chromosome Location:
Chr.14: 24232417 - 24239242 on Build GRCh38
UniGene ID:
Hs.368855
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s27863
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GMPR2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002000.2
NP_001002000.1
10
1405
NM_001002001.2
NP_001002001.1
10
1346
NM_001002002.2
NP_001002002.1
10
1085
NM_001283021.1
NP_001269950.1
10
1664
NM_001283022.1
NP_001269951.1
8
1509
NM_001283023.1
NP_001269952.1
9
1001
NM_016576.4
NP_057660.2
9
1326
NR_104265.1
10
1429
XM_005267740.3
XP_005267797.1
9
1394
XM_005267741.3
XP_005267798.1
9
1453
XM_005267742.3
XP_005267799.1
9
1272
XM_006720165.2
XP_006720228.1
10
1372
XM_017021356.1
XP_016876845.1
10
1588
XM_017021357.1
XP_016876846.1
11
1444
XM_017021358.1
XP_016876847.1
11
1545
XM_017021359.1
XP_016876848.1
11
1188
XM_017021360.1
XP_016876849.1
7
975
GenBank mRNA
AB032903.1
-
9
952
AF086919.1
-
10
1121
AF135159.1
-
10
1291
AF419346.1
-
10
1195
AK226166.1
-
10
1420
BC003053.1
-
8
1458
3546620
BC008021.1
-
9
1274
3537989
BC009832.2
-
10
1003
3958714
BC093039.1
-
10
1144
30347740
BX161436.1
-
10
1326
BX161518.1
-
10
1216
BX247993.1
-
9
924
CR457156.1
-
9
944
Back To Top
More Information
Panther Classification:
Molecular Function -
reductase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleobase metabolic process
purine-containing compound salvage
GMP metabolic process
oxidation-reduction process
GMP reductase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0