Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130716.1 | NP_001124188.1 | 5 | 707 | ||
NM_016619.2 | NP_057703.1 | 5 | 668 | |||
GenBank mRNA | AF208846.1 | - | 5 | 647 | ||
AF275812.1 | - | 362 | ||||
AF279898.1 | - | 362 | ||||
AJ422147.1 | - | 5 | 649 | |||
AK000140.1 | - | 5 | 668 | |||
BC012205.1 | - | 5 | 691 | 3859609 | ||
DQ007338.1 | - | 5 | 630 |