Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016647.2 | NP_057731.1 | 2 | 1725 | ||
XM_005250955.2 | XP_005251012.1 | 2 | 1566 | |||
GenBank mRNA | AF242773.1 | - | 2 | 1658 | ||
AK055140.1 | - | 1 | 1869 | |||
AK075407.1 | - | 2 | 1653 |