Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170795.1 | NP_001164266.1 | 5 | 436 | ||
NM_016085.4 | NP_057169.2 | 5 | 576 | |||
NM_080592.3 | NP_542159.3 | 5 | 756 | |||
GenBank mRNA | AB009017.1 | - | 5 | 449 | ||
AF077037.1 | - | 5 | 438 | |||
AF144055.2 | - | 5 | 566 | |||
AF275744.1 | - | 6 | 430 | |||
AK291894.1 | - | 5 | 566 | |||
AY358968.1 | - | 5 | 429 | |||
BC002846.1 | - | 5 | 424 | |||
BC011006.1 | - | 5 | 430 | |||
BC021237.2 | - | 5 | 536 | 4303513 | ||
BC035850.1 | - | 5 | 422 | 5753421 | ||
BI194121.1 | - | 5 | 577 | |||
BQ420456.1 | - | 5 | 559 | |||
BQ718371.1 | - | 5 | 471 | |||
CF126865.1 | - | 5 | 569 |