Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137664.1 | NP_001131136.1 | 10 | 1591 | ||
NM_016238.2 | NP_057322.2 | 10 | 1591 | |||
XM_005253903.3 | XP_005253960.1 | 9 | 1457 | |||
XM_011538454.2 | XP_011536756.1 | 10 | 1819 | |||
XM_017019425.1 | XP_016874914.1 | 9 | 1366 | |||
XM_017019426.1 | XP_016874915.1 | 10 | 1586 | |||
XM_017019427.1 | XP_016874916.1 | 10 | 2385 | |||
XM_017019428.1 | XP_016874917.1 | 9 | 1474 | |||
GenBank mRNA | AF191340.1 | - | 10 | 1488 | ||
AK021809.1 | - | 10 | 1570 | |||
AL137586.1 | - | 4 | 645 | |||
AY007104.1 | - | 3 | 961 | |||
BC002941.2 | - | 5 | 735 | |||
BC009498.2 | - | 8 | 1161 | |||
BC017316.2 | - | 6 | 937 | |||
BC040130.1 | - | 10 | 1506 | |||
BC098264.1 | - | 10 | 1487 | |||
BC098295.1 | - | 10 | 1487 | |||
BC098362.1 | - | 10 | 1487 | |||
BC099732.1 | - | 10 | 1487 | |||
BC111798.1 | - | 10 | 1487 | |||
BC141848.1 | - | 10 | 1541 | 8860346 | ||
BC148236.2 | - | 10 | 1488 | 40024981 |