Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPCAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282396.1 | NP_001269325.1 | 3 | 344 | ||
NM_001282397.1 | NP_001269326.1 | 2 | 238 | |||
NM_016257.3 | NP_057341.1 | 2 | 463 | |||
XM_017001447.1 | XP_016856936.1 | 2 | 76 | |||
GenBank mRNA | AB001105.1 | - | 2 | 445 | ||
AK294412.1 | - | 2 | 183 | |||
AK294813.1 | - | 2 | 220 | |||
AK312314.1 | - | 2 | 185 | |||
AL136591.1 | - | 2 | 170 | |||
BC030827.1 | - | 2 | 167 | 4811834 | ||
DA107675.1 | - | 3 | 292 | |||
DA220121.1 | - | 3 | 270 | |||
DC333158.1 | - | 2 | 195 | |||
HY156854.1 | - | 2 | 241 |