Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM14C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165258.1 | NP_001158730.1 | 5 | 622 | ||
NM_016462.3 | NP_057546.1 | 5 | 608 | |||
GenBank mRNA | AF151028.1 | - | 5 | 418 | ||
AK223462.1 | - | 5 | 306 | |||
AK309591.1 | - | 5 | 356 | |||
AK310191.1 | - | 5 | 355 | |||
AL702149.1 | - | 5 | 442 | |||
BC002496.2 | - | 5 | 343 | 3050609 | ||
BC010086.2 | - | 5 | 372 | 2907072 | ||
CX784985.1 | - | 5 | 608 |