Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324069.1 | NP_001310998.1 | 4 | 412 | ||
NM_001324071.1 | NP_001311000.1 | 3 | 325 | |||
NM_001324072.1 | NP_001311001.1 | 3 | 191 | |||
NM_001324075.1 | NP_001311004.1 | 3 | 325 | |||
NM_001324077.1 | NP_001311006.1 | 3 | 191 | |||
NM_001799.3 | NP_001790.1 | 3 | 325 | |||
NR_136690.1 | 3 | 325 | ||||
GenBank mRNA | AK026509.1 | - | 3 | 210 | ||
AK308493.1 | - | 3 | 260 | |||
BC000834.2 | - | 3 | 164 | 3455907 | ||
BC005298.1 | - | 3 | 232 | 3997991 | ||
BI520184.1 | - | 3 | 325 | |||
BU660651.1 | - | 3 | 154 | |||
KU177970.1 | - | 3 | 144 | |||
L20320.1 | - | 4 | 319 | |||
X77303.1 | - | 3 | 187 | |||
X77743.1 | - | 3 | 178 | |||
X79193.1 | - | 3 | 234 | |||
Y13120.1 | - | 3 | 247 |