Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015885.3 | NP_056969.2 | 5 | 2046 | ||
XM_005274048.1 | XP_005274105.1 | 5 | 2046 | |||
XM_005274049.1 | XP_005274106.1 | 5 | 2046 | |||
XM_017017918.1 | XP_016873407.1 | 5 | 2046 | |||
GenBank mRNA | AB020631.1 | - | 5 | 1970 | ||
AF046935.1 | - | 5 | 1998 | |||
AK025103.1 | - | 5 | 1982 | |||
AK302606.1 | - | 5 | 2015 | |||
BC065384.1 | - | 5 | 1938 | 6043635 | ||
BC146778.1 | - | 5 | 1970 | 9007228 |