Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014433.2 | NP_001014433.1 | 3 | 462 | ||
NM_001014837.1 | NP_001014837.1 | 3 | 711 | |||
NM_001014838.1 | NP_001014838.1 | 3 | 680 | |||
NM_001014840.1 | NP_001014840.1 | 3 | 590 | |||
NM_015921.2 | NP_057005.1 | 2 | 853 | |||
GenBank mRNA | AF106943.1 | - | 2 | 490 | ||
AF230924.1 | - | 2 | 276 | |||
AK311556.1 | - | 1 | 842 | |||
AL533788.3 | - | 2 | 673 | |||
BC005890.2 | - | 3 | 287 | 3505574 | ||
BC107751.1 | - | 2 | 490 | 6714433 | ||
BU540844.1 | - | 468 | ||||
CB106834.1 | - | 3 | 416 | |||
CN363642.1 | - | 3 | 492 |