Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016024.2 | NP_057108.2 | 6 | 637 | ||
GenBank mRNA | AF078865.1 | - | 5 | 573 | ||
AF151837.1 | - | 5 | 573 | |||
AK292855.1 | - | 6 | 619 | |||
AL050405.1 | - | 6 | 612 | |||
BC033750.1 | - | 6 | 605 | 5210341 | ||
BC040433.1 | - | 6 | 588 |