Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 | 512 | ||
NM_001127400.1 | NP_001120872.1 | 4 | 748 | |||
NM_001127401.1 | NP_001120873.1 | 5 | 809 | |||
NM_016061.2 | NP_057145.1 | 3 | 451 | |||
XM_017004318.1 | XP_016859807.1 | 5 | 1047 | |||
XM_017004319.1 | XP_016859808.1 | 4 | 485 | |||
XM_017004320.1 | XP_016859809.1 | 4 | 986 | |||
XM_017004321.1 | XP_016859810.1 | 4 | 678 | |||
XM_017004322.1 | XP_016859811.1 | 4 | 909 | |||
GenBank mRNA | AB098739.1 | - | 2 | 248 | ||
AF135161.1 | - | 3 | 355 | |||
AF151885.1 | - | 3 | 373 | |||
AF172940.1 | - | 3 | 375 | |||
AK025730.1 | - | 4 | 389 | |||
AK307099.1 | - | 2 | 164 | |||
AK311404.1 | - | 3 | 162 | |||
BC000836.1 | - | 4 | 326 | 3456031 | ||
BC047237.1 | - | 4 | 909 | 5269563 |