Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUC7L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006107.3 | NP_006098.2 | 8 | 890 | ||
NM_016424.4 | NP_057508.2 | 8 | 890 | |||
XM_005257448.1 | XP_005257505.1 | 8 | 858 | |||
XM_005257449.2 | XP_005257506.1 | 8 | 858 | |||
XM_005257452.1 | XP_005257509.1 | 8 | 858 | |||
XM_006721943.2 | XP_006722006.1 | 8 | 637 | |||
GenBank mRNA | AF147357.1 | - | 3 | 256 | ||
AK001925.1 | - | 8 | 824 | |||
AK023672.1 | - | 8 | 852 | |||
AK225980.1 | - | 8 | 858 | |||
AK290540.1 | - | 8 | 857 | |||
AK295983.1 | - | 7 | 759 | |||
BC047043.1 | - | 8 | 839 | |||
DQ013876.1 | - | 8 | 826 |