Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011885.1 | NP_001011885.1 | 3 | 801 | ||
NM_025238.3 | NP_079514.1 | 3 | 801 | |||
GenBank mRNA | AF257241.1 | - | 3 | 680 | ||
AF355402.1 | - | 3 | 681 | |||
AF529369.1 | - | 3 | 598 | |||
AK000731.1 | - | 3 | 665 | |||
AK225355.1 | - | 3 | 665 | |||
AK225362.1 | - | 3 | 665 | |||
AK294410.1 | - | 1 | 53 | |||
AL136853.1 | - | 3 | 681 | |||
AY364253.1 | - | 3 | 598 | |||
BC017464.1 | - | 3 | 628 | |||
BC028097.1 | - | 3 | 701 | 5242418 |