Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPA17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017425.3 | NP_059121.1 | 2 | 218 | ||
XM_011542870.2 | XP_011541172.1 | 2 | 408 | |||
GenBank mRNA | AF334735.1 | - | 2 | 217 | ||
AK307271.1 | - | 2 | 155 | |||
AK311806.1 | - | 2 | 161 | |||
BC019224.1 | - | 2 | 150 | |||
BC032457.2 | - | 1 | 110 | 5171849 | ||
DA511916.1 | - | 2 | 267 | |||
KU257683.1 | - | 1 | 82 |