Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 2 | 743 | ||
NM_001256023.1 | NP_001242952.1 | 2 | 507 | |||
NM_016929.4 | NP_058625.2 | 2 | 507 | |||
XM_011514692.2 | XP_011512994.1 | 2 | 743 | |||
XM_011514694.2 | XP_011512996.1 | 2 | 743 | |||
XM_017010953.1 | XP_016866442.1 | 3 | 988 | |||
GenBank mRNA | AF176920.1 | - | 2 | 306 | ||
AF216941.1 | - | 2 | 457 | |||
AK075144.1 | - | 2 | 446 | |||
AK075163.1 | - | 2 | 358 | |||
AK097048.1 | - | 2 | 743 | |||
AK222544.1 | - | 2 | 475 | |||
AK223130.1 | - | 2 | 478 | |||
AK314416.1 | - | 2 | 323 | |||
AY032602.1 | - | 2 | 637 | |||
BC035968.1 | - | 2 | 475 | 4611102 | ||
DA843541.1 | - | 2 | 507 | |||
DQ679794.1 | - | 2 | 475 |