Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001804.2 | NP_001795.2 | 2 | 617 | ||
GenBank mRNA | AF239666.1 | - | 2 | 602 | ||
BC096251.1 | - | 3 | 234 | |||
BC096252.1 | - | 3 | 280 | 40005955 | ||
BC127122.1 | - | 3 | 280 | 40005957 | ||
U15212.1 | - | 2 | 335 | |||
U51095.1 | - | 2 | 602 |