Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020370.2 | NP_065103.1 | 2 | 827 | ||
XM_011538495.2 | XP_011536797.1 | 2 | 1514 | |||
GenBank mRNA | AF237762.1 | - | 2 | 793 | ||
AF282693.1 | - | 2 | 815 | |||
AK290394.1 | - | 1 | 1361 | |||
BC020614.1 | - | 2 | 825 | 4279185 | ||
FJ348261.1 | - | 1 | 725 |
Set Membership: |
G-protein coupled receptor |