Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIS18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018944.2 | NP_061817.1 | 3 | 526 | ||
XM_017028400.1 | XP_016883889.1 | 3 | 617 | |||
XM_017028401.1 | XP_016883890.1 | 3 | 617 | |||
XM_017028402.1 | XP_016883891.1 | 3 | 225 | |||
GenBank mRNA | AF231921.1 | - | 3 | 498 | ||
AF387845.1 | - | 3 | 496 | |||
AK075281.1 | - | 3 | 498 | |||
AK312073.1 | - | 3 | 617 | |||
BC042917.1 | - | 3 | 502 | 5934336 | ||
BU567844.1 | - | 1 | 93 | |||
BU616573.1 | - | 3 | 529 |