Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCUN1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308101.1 | NP_001295030.1 | 4 | 589 | ||
NM_020640.3 | NP_065691.2 | 4 | 571 | |||
XM_011512912.1 | XP_011511214.1 | 4 | 540 | |||
XM_011512914.1 | XP_011511216.1 | 4 | 579 | |||
XM_011512915.2 | XP_011511217.1 | 5 | 1101 | |||
XM_011512916.2 | XP_011511218.1 | 4 | 1029 | |||
XM_017006633.1 | XP_016862122.1 | 3 | 456 | |||
XM_017006634.1 | XP_016862123.1 | 4 | 545 | |||
XM_017006635.1 | XP_016862124.1 | 5 | 1101 | |||
GenBank mRNA | AF292100.2 | - | 4 | 488 | ||
AF456425.1 | - | 4 | 474 | |||
AF456426.1 | - | 4 | 474 | |||
AK025764.1 | - | 2 | 134 | |||
AK056335.1 | - | 4 | 504 | |||
AK297328.1 | - | 5 | 388 | |||
AK314480.1 | - | 4 | 488 | |||
BC009478.2 | - | 4 | 493 | 3939758 | ||
BC013163.1 | - | 3 | 444 | |||
BI828039.1 | - | 4 | 529 | |||
CD687300.1 | - | 4 | 549 |