Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018992.3 | NP_061865.1 | 2 | 329 | ||
GenBank mRNA | AK000047.1 | - | 2 | 327 | ||
BC007314.2 | - | 2 | 282 | 2989642 | ||
DA134449.1 | - | 2 | 329 |
Gene Symbol: LOC652276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015441.1 | 3 | 957 | |||
GenBank mRNA | AK096394.1 | - | 3 | 991 | ||
AK291606.1 | - | 3 | 941 | |||
BC062576.1 | - | 3 | 921 | |||
BC080650.1 | - | 3 | 887 |