Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284301.2 | NP_001271230.1 | 11, 12 | 2284 | ||
NM_001284302.2 | NP_001271231.1 | 11, 12 | 2281 | |||
NM_018685.4 | NP_061155.2 | 12, 13 | 2395 | |||
XM_006715746.1 | XP_006715809.1 | 12, 13 | 2392 | |||
XM_006715747.3 | XP_006715810.1 | 11, 12 | 2281 | |||
XM_017012354.1 | XP_016867843.1 | 12, 13 | 2380 | |||
XM_017012355.1 | XP_016867844.1 | 12, 13 | 2380 | |||
XM_017012356.1 | XP_016867845.1 | 11, 12 | 2269 | |||
GenBank mRNA | AF273437.1 | - | 12, 13 | 2330 | ||
AK001468.1 | - | 2, 3 | 314 | |||
AK001472.1 | - | 7, 8 | 1062 | |||
AK023208.1 | - | 3, 4 | 436 | |||
AK291254.1 | - | 11, 12 | 2213 | |||
AK299803.1 | - | 12, 13 | 2097 | |||
BC034692.1 | - | 1, 2 | 91 | |||
BC070066.1 | - | 11, 12 | 2266 | 4825802 | ||
CR936650.1 | - | 9, 10 | 1485 |