Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242581.1 | NP_001229510.1 | 12 | 2808 | ||
NM_018993.3 | NP_061866.1 | 11 | 2548 | |||
XM_005260731.2 | XP_005260788.1 | 13 | 3004 | |||
XM_005260733.2 | XP_005260790.1 | 9 | 2356 | |||
XM_006723574.3 | XP_006723637.1 | 14 | 2986 | |||
XM_006723575.3 | XP_006723638.1 | 13 | 2772 | |||
XM_006723577.2 | XP_006723640.1 | 12 | 2621 | |||
XM_011529255.2 | XP_011527557.2 | 12 | 3081 | |||
XM_011529257.1 | XP_011527559.1 | 12 | 2677 | |||
XM_011529258.2 | XP_011527560.1 | 12 | 2689 | |||
XM_011529259.2 | XP_011527561.1 | 11 | 2601 | |||
XM_017027887.1 | XP_016883376.1 | 15 | 3327 | |||
XM_017027888.1 | XP_016883377.1 | 16 | 3399 | |||
XM_017027889.1 | XP_016883378.1 | 11 | 2928 | |||
XM_017027890.1 | XP_016883379.1 | 13 | 2759 | |||
XM_017027891.1 | XP_016883380.1 | 13 | 2797 | |||
XM_017027892.1 | XP_016883381.1 | 11 | 2612 | |||
GenBank mRNA | AB060339.1 | - | 11 | 2512 | ||
AK094884.1 | - | 12 | 2809 | |||
AL136924.1 | - | 10 | 2429 | |||
BC128065.1 | - | 8 | 1249 | 40035872 | ||
M37190.1 | - | 5 | 1240 |