Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001483.2 | NP_001001483.1 | 14 | 1487 | ||
NM_019028.2 | NP_061901.2 | 15 | 1633 | |||
XM_005252996.2 | XP_005253053.1 | 15 | 1605 | |||
XM_011520193.1 | XP_011518495.1 | 12 | 1246 | |||
XM_011520194.2 | XP_011518496.1 | 16 | 1752 | |||
XM_011520195.1 | XP_011518497.1 | 15 | 1543 | |||
GenBank mRNA | AB024495.1 | - | 15 | 1581 | ||
AB097014.1 | - | 15 | 1631 | |||
AB097027.1 | - | 15 | 1631 | |||
AK001714.1 | - | 15 | 1631 | |||
AK001831.1 | - | 10 | 1005 | |||
AK123539.1 | - | 1 | 1334 | |||
AK291348.1 | - | 15 | 1608 | |||
AL833714.1 | - | 11 | 1083 | |||
BC036020.1 | - | 15 | 1618 | 5272445 | ||
BC050690.1 | - | 14 | 1487 | 6171322 | ||
BC056152.1 | - | 15 | 1633 | 6170701 | ||
BX648156.1 | - | 13 | 5499 |