Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPVL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019029.2 | NP_061902.2 | 5 | 659 | ||
NM_031311.3 | NP_112601.3 | 5 | 571 | |||
XM_011515437.1 | XP_011513739.1 | 5 | 738 | |||
XM_017012366.1 | XP_016867855.1 | 5 | 571 | |||
GenBank mRNA | AF106704.2 | - | 5 | 491 | ||
AF217508.1 | - | 4 | 458 | |||
AF282617.1 | - | 5 | 521 | |||
AK075433.1 | - | 5 | 572 | |||
AK124472.1 | - | 9 | 1099 | |||
AK225545.1 | - | 5 | 522 | |||
AY358549.1 | - | 5 | 548 | |||
BC016838.2 | - | 5 | 619 | 3888647 |