Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROBO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301088.1 | NP_001288017.1 | 16 | 2448 | ||
NM_019055.5 | NP_061928.4 | 16 | 2654 | |||
XM_006718861.1 | XP_006718924.1 | 16 | 2473 | |||
XM_011542875.1 | XP_011541177.1 | 10 | 1416 | |||
GenBank mRNA | AF361473.1 | - | 16 | 2583 | ||
AK000805.1 | - | 8 | 1110 | |||
AK025195.1 | - | 4 | 504 | |||
AK027852.1 | - | 16 | 2552 | |||
AK074163.1 | - | 14 | 2338 | |||
AK289769.1 | - | 16 | 2553 | |||
AK297845.1 | - | 10 | 1234 | |||
AK302628.1 | - | 16 | 2318 | |||
AY358083.1 | - | 16 | 2505 | |||
BC014995.1 | - | 15 | 2266 | |||
BC025675.1 | - | 3 | 1977 | |||
BC039602.1 | - | 16 | 2542 | 5590503 | ||
BC064643.1 | - | 6 | 801 | |||
BC111562.1 | - | 16 | 2548 | 40068748 | ||
BC111748.1 | - | 16 | 2548 | 40068752 |