Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286238.1 | NP_001273167.1 | 3, 4 | 439 | ||
NM_019073.3 | NP_061946.1 | 3, 4 | 439 | |||
XM_006710699.2 | XP_006710762.1 | 3, 4 | 433 | |||
XM_006710701.3 | XP_006710764.1 | 3, 4 | 433 | |||
XM_011541606.1 | XP_011539908.1 | 3, 4 | 433 | |||
XM_017001509.1 | XP_016856998.1 | 3, 4 | 433 | |||
XM_017001511.1 | XP_016857000.1 | 3, 4 | 433 | |||
XM_017001513.1 | XP_016857002.1 | 1, 2 | 30 | |||
XM_017001514.1 | XP_016857003.1 | 3, 4 | 260 | |||
XM_017001515.1 | XP_016857004.1 | 3, 4 | 245 | |||
GenBank mRNA | AK000869.1 | - | 3, 4 | 404 | ||
BC020660.1 | - | 3, 4 | 241 | 4657425 | ||
DB025426.1 | - | 3, 4 | 435 |