Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040450.2 | NP_001035540.1 | 5 | 1377 | ||
NM_001040453.2 | NP_001035543.1 | 5 | 1207 | |||
XM_011521687.1 | XP_011519989.1 | 6 | 1429 | |||
XM_011521688.1 | XP_011519990.1 | 6 | 1429 | |||
XM_011521689.1 | XP_011519991.1 | 6 | 1327 | |||
XM_011521690.1 | XP_011519992.1 | 5 | 1273 | |||
GenBank mRNA | AB032990.1 | - | 5 | 484 | ||
AK075319.1 | - | 5 | 1255 | |||
AK297291.1 | - | 5 | 1255 | |||
AL833610.1 | - | 5 | 1378 | |||
BC140811.1 | - | 5 | 1207 | 9021679 | ||
BC144616.1 | - | 5 | 1207 | 9053147 |