Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286772.1 | NP_001273701.1 | 8, 9 | 853 | ||
NM_018201.4 | NP_060671.3 | 10, 11 | 1228 | |||
XM_005252060.2 | XP_005252117.1 | 9, 10 | 1131 | |||
GenBank mRNA | AB209565.1 | - | 7, 8 | 947 | ||
AB449899.1 | - | 10, 11 | 1081 | |||
AK001605.1 | - | 8, 9 | 711 | |||
AK027457.1 | - | 2, 3 | 272 | |||
AK123828.1 | - | 6, 7 | 842 | |||
AK289777.1 | - | 10, 11 | 1140 | |||
AK295183.1 | - | 7, 8 | 931 | |||
AK307556.1 | - | 6, 7 | 830 | |||
BC014898.1 | - | 10, 11 | 1103 | |||
BC132817.1 | - | 10, 11 | 1101 | 40146839 | ||
BC136509.1 | - | 10, 11 | 1101 | 9020499 | ||
BC150311.1 | - | 10, 11 | 1137 | 8860447 |