Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127458.1 | NP_001120930.1 | 2 | 181 | ||
NM_001323561.1 | NP_001310490.1 | 2 | 189 | |||
NM_001323562.1 | NP_001310491.1 | 2 | 196 | |||
NM_001323563.1 | NP_001310492.1 | 2 | 564 | |||
NM_001323564.1 | NP_001310493.1 | 2 | 190 | |||
NM_019095.5 | NP_061968.1 | 2 | 502 | |||
NR_136617.1 | 2 | 502 | ||||
XM_005260738.4 | XP_005260795.1 | 2 | 166 | |||
XM_011529263.1 | XP_011527565.1 | 2 | 477 | |||
GenBank mRNA | AF241784.1 | - | 2 | 421 | ||
AK310368.1 | - | 2 | 166 | |||
AL832419.1 | - | 2 | 249 | |||
BC069010.1 | - | 2 | 425 | 5092704 | ||
BM563835.1 | - | 2 | 184 | |||
BM763234.1 | - | 2 | 482 | |||
BP368984.1 | - | 2 | 181 | |||
BU852955.1 | - | 3 | 209 | |||
DQ386730.1 | - | 2 | 328 |