Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161779.1 | NP_001155251.1 | 3 | 1279 | ||
NM_001161780.1 | NP_001155252.1 | 3 | 1153 | |||
NM_001161781.1 | NP_001155253.1 | 2 | 1140 | |||
NM_018444.3 | NP_060914.2 | 2 | 1197 | |||
XM_011517135.2 | XP_011515437.1 | 3 | 1050 | |||
XM_011517136.1 | XP_011515438.1 | 3 | 1157 | |||
XM_017013587.1 | XP_016869076.1 | 2 | 1212 | |||
XM_017013588.1 | XP_016869077.1 | 3 | 1120 | |||
GenBank mRNA | AF155661.1 | - | 3 | 1167 | ||
AK057079.1 | - | 2 | 1109 | |||
AK126862.1 | - | 1 | 1574 | |||
AK302651.1 | - | 2 | 1083 | |||
BC047619.1 | - | 2 | 1118 | 4824168 | ||
BC064978.1 | - | 3 | 1137 | |||
BC098343.1 | - | 1 | 951 | 40010016 |