Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017824.4 | NP_060294.1 | 4 | 823 | ||
XM_011539886.2 | XP_011538188.1 | 5 | 971 | |||
GenBank mRNA | AB191202.1 | - | 4 | 579 | ||
AK000452.1 | - | 4 | 825 | |||
AK294799.1 | - | 4 | 897 | |||
AK309175.1 | - | 3 | 692 | |||
BC015480.1 | - | 4 | 783 | 3905766 |