Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017521.2 | NP_059991.1 | 3 | 1647 | ||
GenBank mRNA | BC023511.2 | - | 3 | 1542 | 4130242 | |
Y08976.1 | - | 3 | 1649 |