Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017583.5 | NP_060053.2 | 3 | 1324 | ||
GenBank mRNA | AK021823.1 | - | 3 | 430 | ||
AK074242.1 | - | 3 | 1148 | |||
BC013166.1 | - | 3 | 1271 | 3924490 | ||
BC024031.2 | - | 3 | 1171 | 3532065 |