Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 55 | 7572 | ||
GenBank mRNA | AL122111.1 | - | 12 | 1073 | ||
BC034223.1 | - | 17 | 2040 | 4838774 |