Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 58 | 7840 | ||
GenBank mRNA | AK128150.1 | - | 1 | 2447 | ||
AL122111.1 | - | 15 | 1341 | |||
BC034223.1 | - | 20 | 2308 | 4838774 |