Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BORCS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017622.2 | NP_060092.2 | 1 | 744 | ||
GenBank mRNA | AK000021.1 | - | 1 | 570 | ||
AK222506.1 | - | 1 | 570 | |||
BC009261.2 | - | 1 | 527 | 3928189 | ||
BC018880.2 | - | 1 | 626 | 3615792 | ||
BC070272.1 | - | 1 | 641 | 4672989 | ||
DR006664.1 | - | 1 | 363 |